Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,2-dimethoxypropane, 97%, ACROS Organics™

Click to view available options
Quantity:
10mL
Packaging:
Glass bottle
126-38-5
C5H11BrO2
183.045
MFCD00000208
SGTITUFGCGGICE-UHFFFAOYSA-N
propane,1-bromo-2,2-dimethoxy,propane, 1-bromo-2,2-dimethoxy,bromoacetone dimethyl acetal,bromo-2,2dimethoxypropane,bromo-2,2-dimethoxypropane,1-bromo-2,2-dimethoxypropan,1-bromo-2,2-dimethoxyprpopane,1-bromo-2,2-dimethoxy propane,1-bromo-2,2-dimethoxy-propane,bromoacetone dimethyl ketal
67175
1-bromo-2,2-dimethoxypropane
CC(CBr)(OC)OC
126-38-5
C5H11BrO2
183.045
MFCD00000208
SGTITUFGCGGICE-UHFFFAOYSA-N
propane,1-bromo-2,2-dimethoxy,propane, 1-bromo-2,2-dimethoxy,bromoacetone dimethyl acetal,bromo-2,2dimethoxypropane,bromo-2,2-dimethoxypropane,1-bromo-2,2-dimethoxypropan,1-bromo-2,2-dimethoxyprpopane,1-bromo-2,2-dimethoxy propane,1-bromo-2,2-dimethoxy-propane,bromoacetone dimethyl ketal
67175
1-bromo-2,2-dimethoxypropane
CC(CBr)(OC)OC
Color Yellow
Quantity 10mL
Refractive Index 1.4465 to 1.4485 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C(OCH3)2CH2Br
Formula Weight 183.04
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1-Bromo-2, 2-dimethoxypropane, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-784-7

TSCA : TSCA