Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
54149-17-6
C5H11BrO2
183.04
MFCD00000238
HUXJXNSHCKHFIL-UHFFFAOYSA-N
1-bromo-2-2-methoxyethoxy ethane,1-2-bromoethoxy-2-methoxyethane,1-bromo-3,6-dioxaheptane,ethane, 1-2-bromoethoxy-2-methoxy,2-2-methoxyethoxy ethyl bromide,1-2-bromoethoxy-2-methoxy-ethane,1-bromo-2-2-methoxy-ethoxy ethane,mpeg2-br,acmc-1alws,7-bromo-2,5-dioxaheptane
123532
1-(2-bromoethoxy)-2-methoxyethane
COCCOCCBr
54149-17-6
C5H11BrO2
183.04
MFCD00000238
HUXJXNSHCKHFIL-UHFFFAOYSA-N
1-bromo-2-2-methoxyethoxy ethane,1-2-bromoethoxy-2-methoxyethane,1-bromo-3,6-dioxaheptane,ethane, 1-2-bromoethoxy-2-methoxy,2-2-methoxyethoxy ethyl bromide,1-2-bromoethoxy-2-methoxy-ethane,1-bromo-2-2-methoxy-ethoxy ethane,mpeg2-br,acmc-1alws,7-bromo-2,5-dioxaheptane
123532
1-(2-bromoethoxy)-2-methoxyethane
COCCOCCBr
Quantity 50g
Refractive Index 1.455
Specific Gravity 1.347
Packaging Glass bottle
Stabilizer 0.1% sodium carbonate
Color Colorless to Yellow
Infrared Spectrum Authentic
Flash Point 73°C
Density 1.3470g/mL
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula CH3(OCH2CH2)2Br
Formula Weight 183.04
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material 1-Bromo-2-(2-methoxyethoxy)ethane, 90%
Show More Show Less

EINECSNumber : 259-000-6

Recommended Storage : Refrigerator +4°C