Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2,3,5,6-tetrafluorobenzene, 99%

Click to view available options
Quantity:
1g
5g
25g
1559-88-2
C6HBrF4
228.972
MFCD00000308
YHAFCGSUIAFUCX-UHFFFAOYSA-N
1-bromo-2,3,5,6-tetrafluorobenzene,2,3,5,6-tetrafluorobromobenzene,bromo-2,3,5,6-tetrafluorobenzene,1-bromo-2,3,5,6-tetrafluorbenzene,benzene, 3-bromo-1,2,4,5-tetrafluoro,2,3,5,6-tetrafluoro bromobenzene,2,3,5,6-tetrafluorobromo benzene,1,2,4,5-tetrafluoro-3-bromobenzene,pubchem2196,acmc-1bwvg
73794
3-bromo-1,2,4,5-tetrafluorobenzene
C1=C(C(=C(C(=C1F)F)Br)F)F
1559-88-2
C6HBrF4
228.972
MFCD00000308
YHAFCGSUIAFUCX-UHFFFAOYSA-N
1-bromo-2,3,5,6-tetrafluorobenzene,2,3,5,6-tetrafluorobromobenzene,bromo-2,3,5,6-tetrafluorobenzene,1-bromo-2,3,5,6-tetrafluorbenzene,benzene, 3-bromo-1,2,4,5-tetrafluoro,2,3,5,6-tetrafluoro bromobenzene,2,3,5,6-tetrafluorobromo benzene,1,2,4,5-tetrafluoro-3-bromobenzene,pubchem2196,acmc-1bwvg
73794
3-bromo-1,2,4,5-tetrafluorobenzene
C1=C(C(=C(C(=C1F)F)Br)F)F
Applications
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Flash Point 53°C (129°F)
Boiling Point 142°C to 144°C
Quantity 1g
Odor Aromatic
UN Number UN1993
Formula Weight 228.98
Percent Purity 99%
Chemical Name or Material 1-Bromo-2,3,5,6-tetrafluorobenzene

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.