Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,3,5,6-tetrafluorobenzene, 99%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1559-88-2
C6HBrF4
228.972
YHAFCGSUIAFUCX-UHFFFAOYSA-N
1-bromo-2,3,5,6-tetrafluorobenzene,2,3,5,6-tetrafluorobromobenzene,bromo-2,3,5,6-tetrafluorobenzene,1-bromo-2,3,5,6-tetrafluorbenzene,benzene, 3-bromo-1,2,4,5-tetrafluoro,2,3,5,6-tetrafluoro bromobenzene,2,3,5,6-tetrafluorobromo benzene,1,2,4,5-tetrafluoro-3-bromobenzene,pubchem2196,acmc-1bwvg
73794
3-bromo-1,2,4,5-tetrafluorobenzene
C1=C(C(=C(C(=C1F)F)Br)F)F
1559-88-2
C6HBrF4
228.972
YHAFCGSUIAFUCX-UHFFFAOYSA-N
1-bromo-2,3,5,6-tetrafluorobenzene,2,3,5,6-tetrafluorobromobenzene,bromo-2,3,5,6-tetrafluorobenzene,1-bromo-2,3,5,6-tetrafluorbenzene,benzene, 3-bromo-1,2,4,5-tetrafluoro,2,3,5,6-tetrafluoro bromobenzene,2,3,5,6-tetrafluorobromo benzene,1,2,4,5-tetrafluoro-3-bromobenzene,pubchem2196,acmc-1bwvg
73794
3-bromo-1,2,4,5-tetrafluorobenzene
C1=C(C(=C(C(=C1F)F)Br)F)F
Quantity 1g
Packaging Glass bottle
Assay Percent Range 99%
Formula Weight 228.97
Chemical Name or Material 1-Bromo-2,3,5,6-tetrafluorobenzene