Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2,3-dichlorobenzene, 98%

Click to view available options
Quantity:
25g
100g
56961-77-4
C6H3BrCl2
225.894
MFCD00000536
HVKCZUVMQPUWSX-UHFFFAOYSA-N
2,3-dichlorobromobenzene,benzene, 1-bromo-2,3-dichloro,benzene, bromodichloro,2,3-dichloro-1-bromobenzene,1-bromo-2,3-dichloro-benzene,attercop-chm at113305,bromodichlorobenzene,pubchem3607,monobromodichlorobenzene,acmc-209lv2
42066
1-bromo-2,3-dichlorobenzene
C1=CC(=C(C(=C1)Br)Cl)Cl
56961-77-4
C6H3BrCl2
225.894
MFCD00000536
HVKCZUVMQPUWSX-UHFFFAOYSA-N
2,3-dichlorobromobenzene,benzene, 1-bromo-2,3-dichloro,benzene, bromodichloro,2,3-dichloro-1-bromobenzene,1-bromo-2,3-dichloro-benzene,attercop-chm at113305,bromodichlorobenzene,pubchem3607,monobromodichlorobenzene,acmc-209lv2
42066
1-bromo-2,3-dichlorobenzene
C1=CC(=C(C(=C1)Br)Cl)Cl
Boiling Point 243°C
Quantity 100g
Melting Point 56°C to 60°C
Beilstein 2246806
Formula Weight 225.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,3-dichlorobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning