Alfa Aesar™ 1-Bromo-2,4,5-trifluorobenzene, 98+%

Manufacturer: Alfa Aesar™ A1196206

Quantity 5g

 View more versions of this product

Catalog No. AAA1196206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 327-52-6
    Molecular Formula C6H2BrF3
    Formula Weight 210.98
    MDL Number MFCD00000306
    UN Number UN1993
    Beilstein 1636588
    Synonym 2,4,5-Trifluorobromobenzene
    Chemical Name or Material 1-Bromo-2,4,5-trifluorobenzene

    Specifications

    Formula Weight 210.98
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    CAS 327-52-6
    UN Number UN1993
    Melting Point −19°C
    Beilstein 1636588
    Synonym 2,4,5-Trifluorobromobenzene
    Flash Point 55°C (131°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C6H2BrF3
    MDL Number MFCD00000306
    Boiling Point 143° to 144°C
    Chemical Name or Material 1-Bromo-2,4,5-trifluorobenzene
    Assay ≥98%

    Description

    Applications
    1-Bromo-2,4,5-trifluorobenzene is the intermediate of liquid crystals. The derivatives of this compound can be used as intermediates for the synthesis of qunolone antibacterial. It is employed in the synthesis of 3-ethenyl, 3-ethynyl, 3-aryl, and 3-cyclopropyl-2, 4, 5-trifluorobenzoic acids from 1-bromo-2, 4, 5-trifluorobenzene and 2, 4, 5-trifluoro-3-hydroxybenzoic acid.

    Solubility
    Insoluble in water.

    Notes
    Store at room temperature. Keep away from oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit