Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2,4,5-trifluorobenzene, 98+%

Manufacturer:  Alfa Aesar™ A1196214

Quantity 25g

 View more versions of this product

Catalog No. AAA1196214


  Check Availability
Add to cart

Specifications

Description

Applications
1-Bromo-2,4,5-trifluorobenzene is the intermediate of liquid crystals. The derivatives of this compound can be used as intermediates for the synthesis of qunolone antibacterial. It is employed in the synthesis of 3-ethenyl, 3-ethynyl, 3-aryl, and 3-cyclopropyl-2, 4, 5-trifluorobenzoic acids from 1-bromo-2, 4, 5-trifluorobenzene and 2, 4, 5-trifluoro-3-hydroxybenzoic acid.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

327-52-6
210.98
UN1993
2,4,5-Trifluorobromobenzene
C6H2BrF3
MFCD00000306
1636588
1-Bromo-2,4,5-trifluorobenzene

Specifications

327-52-6
210.98
UN1993
1636588
143° to 144°C
55°C (131°F)
≥98%
C6H2BrF3
MFCD00000306
−19°C
2,4,5-Trifluorobromobenzene
25g
1-Bromo-2,4,5-trifluorobenzene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit