Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,4-dimethoxybenzene 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
17715-69-4
C8H9BrO2
217.06
MFCD00009844
NIUZVSQOXJIHBL-UHFFFAOYSA-N
2,4-dimethoxybromobenzene,1,3-dimethoxy-4-bromobenzene,4-bromo-3-methoxyanisole,benzene, 1-bromo-2,4-dimethoxy,4-bromoresorcinol dimethyl ether,4-bromoresocinol dimethyl ether,4-bromo-1,3-dimethoxybenzene,1-brom-2,4-dimethoxybenzol,pubchem3036,acmc-1c5dw
87266
1-bromo-2,4-dimethoxybenzene
COC1=CC(=C(C=C1)Br)OC
17715-69-4
C8H9BrO2
217.06
MFCD00009844
NIUZVSQOXJIHBL-UHFFFAOYSA-N
2,4-dimethoxybromobenzene,1,3-dimethoxy-4-bromobenzene,4-bromo-3-methoxyanisole,benzene, 1-bromo-2,4-dimethoxy,4-bromoresorcinol dimethyl ether,4-bromoresocinol dimethyl ether,4-bromo-1,3-dimethoxybenzene,1-brom-2,4-dimethoxybenzol,pubchem3036,acmc-1c5dw
87266
1-bromo-2,4-dimethoxybenzene
COC1=CC(=C(C=C1)Br)OC
Melting Point 25°C to 26°C
Boiling Point 153°C to 155°C (18.0mmHg)
Density 1.5070g/mL
Flash Point 109°C
Specific Gravity 1.507
Packaging Glass bottle
Refractive Index 1.5718 to 1.5738
Color Yellow
Infrared Spectrum Authentic
Quantity 5mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H3(OCH3)Br
Beilstein 06, II, 819
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 217.06
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Bromo-2, 4-dimethoxybenzene, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 241-717-0

TSCA : TSCA