Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2,4-dimethyl-5-nitrobenzene, 95%

Click to view available options
Quantity:
1g
5g
25g
69383-59-1
C8H8BrNO2
230.061
MFCD00075824
HYYJANCKMGXHLQ-UHFFFAOYSA-N
5-bromo-2,4-dimethylnitrobenzene,4-bromo-6-nitro-m-xylene,benzene, 1-bromo-2,4-dimethyl-5-nitro,1-bromo-2,4-dimethyl-5-nitro-benzene,2-nitro-4-bromo-m-xylene,2,4-dimethyl-5-nitro-1-bromobenzene,pubchem19926,benzene,1-bromo-2,4-dimethyl-5-nitro
12474326
1-bromo-2,4-dimethyl-5-nitrobenzene
CC1=CC(=C(C=C1[N+](=O)[O-])Br)C
69383-59-1
C8H8BrNO2
230.061
MFCD00075824
HYYJANCKMGXHLQ-UHFFFAOYSA-N
5-bromo-2,4-dimethylnitrobenzene,4-bromo-6-nitro-m-xylene,benzene, 1-bromo-2,4-dimethyl-5-nitro,1-bromo-2,4-dimethyl-5-nitro-benzene,2-nitro-4-bromo-m-xylene,2,4-dimethyl-5-nitro-1-bromobenzene,pubchem19926,benzene,1-bromo-2,4-dimethyl-5-nitro
12474326
1-bromo-2,4-dimethyl-5-nitrobenzene
CC1=CC(=C(C=C1[N+](=O)[O-])Br)C
Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Color Yellow
Quantity 5g
UN Number UN2811
Beilstein 1948547
Formula Weight 230.06
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 1-Bromo-2,4-dimethyl-5-nitrobenzene

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C