Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2-chloro-1,1,2-trifluoro-3-butene, 97%

Click to view available options
Quantity:
5g
25g
374-25-4
C4H3BrClF3
223.417
MFCD00039225
VULPFOSLGWWARI-UHFFFAOYSA-N
1-bromo-2-chloro-1,1,2-trifluoro-3-butene,4-bromo-3-chloro-3,4,4-trifluorobutene-1,1-butene, 4-bromo-3-chloro-3,4,4-trifluoro,4-bromo-3-chloro-3,4,4-trifluorobutene,4-bromo-3-chloro-3,4,4-trifluoro-1-butene,acmc-1ahj4,1-butene,4,4-trifluoro,br-cf2-cfcl-ch=ch2,1-bromo-2-chloro-1,1,2-trifluorobut-3-ene,1-butene,4-bromo-3-chloro-3,4,4-trifluoro
136217
4-bromo-3-chloro-3,4,4-trifluorobut-1-ene
C=CC(C(F)(F)Br)(F)Cl
374-25-4
C4H3BrClF3
223.417
MFCD00039225
VULPFOSLGWWARI-UHFFFAOYSA-N
1-bromo-2-chloro-1,1,2-trifluoro-3-butene,4-bromo-3-chloro-3,4,4-trifluorobutene-1,1-butene, 4-bromo-3-chloro-3,4,4-trifluoro,4-bromo-3-chloro-3,4,4-trifluorobutene,4-bromo-3-chloro-3,4,4-trifluoro-1-butene,acmc-1ahj4,1-butene,4,4-trifluoro,br-cf2-cfcl-ch=ch2,1-bromo-2-chloro-1,1,2-trifluorobut-3-ene,1-butene,4-bromo-3-chloro-3,4,4-trifluoro
136217
4-bromo-3-chloro-3,4,4-trifluorobut-1-ene
C=CC(C(F)(F)Br)(F)Cl
Boiling Point 98°C to 99°C
Quantity 25g
Beilstein 1758700
Formula Weight 223.43
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-chloro-1,1,2-trifluoro-3-butene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning