Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2-(difluoromethoxy)benzene, 97+%

Click to view available options
Quantity:
2g
10g
175278-33-8
C7H5BrF2O
223.017
MFCD00221468
VHEICCMNYWNFQX-UHFFFAOYSA-N
1-bromo-2-difluoromethoxy benzene,2-difluoromethoxy bromobenzene,benzene, 1-bromo-2-difluoromethoxy,2-bromophenyl difluoromethyl ether,2-bromo-alpha,alpha-difluoroanisole,1-difluoromethoxy-2-bromobenzene,acmc-209e9t,ksc493k0n,1-bromo-2-difluoromethoxy-benzene,attercop-chm at123241
2737004
1-bromo-2-(difluoromethoxy)benzene
C1=CC=C(C(=C1)OC(F)F)Br
175278-33-8
C7H5BrF2O
223.017
MFCD00221468
VHEICCMNYWNFQX-UHFFFAOYSA-N
1-bromo-2-difluoromethoxy benzene,2-difluoromethoxy bromobenzene,benzene, 1-bromo-2-difluoromethoxy,2-bromophenyl difluoromethyl ether,2-bromo-alpha,alpha-difluoroanisole,1-difluoromethoxy-2-bromobenzene,acmc-209e9t,ksc493k0n,1-bromo-2-difluoromethoxy-benzene,attercop-chm at123241
2737004
1-bromo-2-(difluoromethoxy)benzene
C1=CC=C(C(=C1)OC(F)F)Br
Quantity 10g
Boiling Point 85°C to 87°C (15mmHg)
Formula Weight 223.02
Percent Purity ≥97%
Chemical Name or Material 1-Bromo-2-(difluoromethoxy)benzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning