Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene, 98%

Click to view available options
Quantity:
1g
5g
25g
168971-68-4
C7H3BrF4O
258.998
MFCD06657548
KZMHSGBETSENAT-UHFFFAOYSA-N
1-bromo-2-fluoro-4-trifluoromethoxy benzene,4-bromo-3-fluoro trifluoromethoxy benzene,3-fluoro-4-bromobenzenetrifluoromethoxy,4-bromo-3-fluoro-trifluoromethoxy benzene,benzene, 1-bromo-2-fluoro-4-trifluoromethoxy,pubchem1075,acmc-1c50f,ksc498c3b,4-bromo-3-fluorotrifluoromethoxy-benzene,2-fluoro-4-trifluoromethoxy bromobenzene
22065211
1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
C1=CC(=C(C=C1OC(F)(F)F)F)Br
168971-68-4
C7H3BrF4O
258.998
MFCD06657548
KZMHSGBETSENAT-UHFFFAOYSA-N
1-bromo-2-fluoro-4-trifluoromethoxy benzene,4-bromo-3-fluoro trifluoromethoxy benzene,3-fluoro-4-bromobenzenetrifluoromethoxy,4-bromo-3-fluoro-trifluoromethoxy benzene,benzene, 1-bromo-2-fluoro-4-trifluoromethoxy,pubchem1075,acmc-1c50f,ksc498c3b,4-bromo-3-fluorotrifluoromethoxy-benzene,2-fluoro-4-trifluoromethoxy bromobenzene
22065211
1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
C1=CC(=C(C=C1OC(F)(F)F)F)Br
Quantity 5g
Color Yellow
Formula Weight 259
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures