Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper

Click to view available options
Quantity:
5g
25g
100g
421-70-5
C2BrF4I
306.824
MFCD00042136
ZYNPYKGTNSXKPI-UHFFFAOYSA-N
1-bromo-2-iodotetrafluoroethane,1-bromotetrafluoro-2-iodoethane,unii-e75xu1n0mr,e75xu1n0mr,ethane, 1-bromo-1,1,2,2-tetrafluoro-2-iodo,ccris 9437,acmc-1amr3,1-bromo-2-iodoperfluoroethane,1-iodo-2-bromotetrafluoroethane,2-iodo-1-bromotetrafluoroethane
67902
1-bromo-1,1,2,2-tetrafluoro-2-iodoethane
C(C(F)(F)I)(F)(F)Br
421-70-5
C2BrF4I
306.824
MFCD00042136
ZYNPYKGTNSXKPI-UHFFFAOYSA-N
1-bromo-2-iodotetrafluoroethane,1-bromotetrafluoro-2-iodoethane,unii-e75xu1n0mr,e75xu1n0mr,ethane, 1-bromo-1,1,2,2-tetrafluoro-2-iodo,ccris 9437,acmc-1amr3,1-bromo-2-iodoperfluoroethane,1-iodo-2-bromotetrafluoroethane,2-iodo-1-bromotetrafluoroethane
67902
1-bromo-1,1,2,2-tetrafluoro-2-iodoethane
C(C(F)(F)I)(F)(F)Br
Boiling Point 78°C
Quantity 5g
Beilstein 1688466
Formula Weight 306.82
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-iodotetrafluoroethane, stabilized with copper

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C