Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-methylnaphthalene, 90%, technical, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2586-62-1
C11H9Br
221.1
MFCD00003871
CMIMBQIBIZZZHQ-UHFFFAOYSA-N
naphthalene, 1-bromo-2-methyl,2-methyl-1-bromonaphthalene,.beta.-methyl-.alpha.-bromonaphthalene,zlchem 421,1-bromo-2-methylnaphtalene,acmc-1cq62,1-bromo-2-methyl-naphthalene,1-bromo-2-methyl naphthalene,1-bromanyl-2-methyl-naphthalene,1-bromonaphthalen-2-yl methyl
75754
1-bromo-2-methylnaphthalene
CC1=C(C2=CC=CC=C2C=C1)Br
2586-62-1
C11H9Br
221.1
MFCD00003871
CMIMBQIBIZZZHQ-UHFFFAOYSA-N
naphthalene, 1-bromo-2-methyl,2-methyl-1-bromonaphthalene,.beta.-methyl-.alpha.-bromonaphthalene,zlchem 421,1-bromo-2-methylnaphtalene,acmc-1cq62,1-bromo-2-methyl-naphthalene,1-bromo-2-methyl naphthalene,1-bromanyl-2-methyl-naphthalene,1-bromonaphthalen-2-yl methyl
75754
1-bromo-2-methylnaphthalene
CC1=C(C2=CC=CC=C2C=C1)Br
Quantity 100g
Flash Point >110°C
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Gravity 1.41
Refractive Index 1.6480 to 1.6500
Density 1.4100g/mL
Boiling Point 296.0°C
Color Yellow
CAS Min % 90.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula CH3C10H6Br
Beilstein 05, 568
Formula Weight 221.1
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Bromo-2-methylnaphthalene, 90-95%
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 219-966-1

TSCA : TSCA