Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-pentyne, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
16400-32-1
C5H7Br
147.01
MFCD00236363
VDHGRVFJBGRHMD-UHFFFAOYSA-N
1-bromo-2-pentyne,2-pentynyl bromide,2-pentyne, 1-bromo,1-bromo-pent-2-yne,1-bromo-2-pentine,2-pentyn-1-yl bromide,acmc-1c6g2
85399
1-bromopent-2-yne
CCC#CCBr
16400-32-1
C5H7Br
147.01
MFCD00236363
VDHGRVFJBGRHMD-UHFFFAOYSA-N
1-bromo-2-pentyne,2-pentynyl bromide,2-pentyne, 1-bromo,1-bromo-pent-2-yne,1-bromo-2-pentine,2-pentyn-1-yl bromide,acmc-1c6g2
85399
1-bromopent-2-yne
CCC#CCBr
Quantity 10g
Boiling Point 40°C to 41°C (20.0mmHg)
Refractive Index 1.508
Packaging Glass bottle
Flash Point 36°C
Color Yellow
Assay Percent Range 96% min. (GC)
Formula Weight 147.01
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-pentyne
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair):

GHS Signal Word: Warning

EINECSNumber : 240-451-2