Alfa Aesar™ 1-Bromo-3,3-diphenylpropane, 99%

Manufacturer: Alfa Aesar™ B2555206

Quantity 5g

 View more versions of this product

Catalog No. AAB2555206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20017-68-9
    Molecular Formula C15H15Br
    Formula Weight 275.18
    MDL Number MFCD00044829
    Chemical Name or Material 1-Bromo-3,3-diphenylpropane

    Specifications

    Formula Weight 275.18
    Molecular Formula C15H15Br
    MDL Number MFCD00044829
    CAS 20017-68-9
    Melting Point 39° to 42°C
    Quantity 5g
    Odor Odorless
    Chemical Name or Material 1-Bromo-3,3-diphenylpropane
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit