Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-3,4,5-trichlorobenzene, 98%

Click to view available options
Quantity:
5g
25g
100g
21928-51-8
C6H2BrCl3
260.336
MFCD00155009
VZUMVBQMJFFYRM-UHFFFAOYSA-N
3,4,5-trichlorobromobenzene,benzene, 5-bromo-1,2,3-trichloro,1-bromo-3,4,5-trichlorobenzene,1-bromo-3,4,5-trichloro-benzene,pubchem23062,acmc-209fpm,ksc497i6l,1,2,3-trichloro-5-bromobenzene
4124400
5-bromo-1,2,3-trichlorobenzene
C1=C(C=C(C(=C1Cl)Cl)Cl)Br
21928-51-8
C6H2BrCl3
260.336
MFCD00155009
VZUMVBQMJFFYRM-UHFFFAOYSA-N
3,4,5-trichlorobromobenzene,benzene, 5-bromo-1,2,3-trichloro,1-bromo-3,4,5-trichlorobenzene,1-bromo-3,4,5-trichloro-benzene,pubchem23062,acmc-209fpm,ksc497i6l,1,2,3-trichloro-5-bromobenzene
4124400
5-bromo-1,2,3-trichlorobenzene
C1=C(C=C(C(=C1Cl)Cl)Cl)Br
Applications
It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Color White
Quantity 5g
Beilstein 2089937
Formula Weight 260.34
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1-Bromo-3,4,5-trichlorobenzene

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning