Alfa Aesar™ 1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%

Manufacturer: Alfa Aesar™ A1233730

Quantity 250g

 View more versions of this product

Catalog No. AAA1233730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 328-70-1
    Molecular Formula C8H3BrF6
    Formula Weight 293.01
    MDL Number MFCD00000381
    Beilstein 2123669
    Synonym 3,5-Bis(trifluoromethyl)bromobenzene; 3,5-Bis(trifluoromethyl)bromobenzene
    Chemical Name or Material 1-Bromo-3,5-bis (trifluoromethyl)benzene

    Specifications

    Formula Weight 293.01
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H3BrF6
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00000381
    Boiling Point 152° to 155°C
    Quantity 250g
    CAS 328-70-1
    Melting Point −16°C
    Beilstein 2123669
    Flash Point >110°C (230°F)
    Health Hazard 1 Warning
    Synonym 3,5-Bis(trifluoromethyl)bromobenzene; 3,5-Bis(trifluoromethyl)bromobenzene
    Chemical Name or Material 1-Bromo-3,5-bis (trifluoromethyl)benzene
    Assay 98%

    Description

    Applications
    1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit