Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,5-dichlorobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
19752-55-7
C6H3BrCl2
225.9
MFCD00000584
DZHFFMWJXJBBRG-UHFFFAOYSA-N
3,5-dichlorobromobenzene,benzene, 1-bromo-3,5-dichloro,1-bromo-3,5-dichloro-benzene,3,5-dichloro bromobenzene,3,5-dichloro-1-bromobenzene,attercop-chm at113311,maybridge1_000881,pubchem3612,acmc-209f1a
29766
1-bromo-3,5-dichlorobenzene
C1=C(C=C(C=C1Cl)Br)Cl
19752-55-7
C6H3BrCl2
225.9
MFCD00000584
DZHFFMWJXJBBRG-UHFFFAOYSA-N
3,5-dichlorobromobenzene,benzene, 1-bromo-3,5-dichloro,1-bromo-3,5-dichloro-benzene,3,5-dichloro bromobenzene,3,5-dichloro-1-bromobenzene,attercop-chm at113311,maybridge1_000881,pubchem3612,acmc-209f1a
29766
1-bromo-3,5-dichlorobenzene
C1=C(C=C(C=C1Cl)Br)Cl
Flash Point >110°C
Boiling Point 232°C
Melting Point 74°C to 77°C
Infrared Spectrum Authentic
Quantity 5g
Color Undesignated
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 225.9
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Bromo-3, 5-dichlorobenzene, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-270-7

TSCA : TSCA