Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,5-dimethoxybenzene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
20469-65-2
C8H9BrO2
217.06
KRWRFIMBWRVMKE-UHFFFAOYSA-N
3,5-dimethoxybromobenzene,benzene, 1-bromo-3,5-dimethoxy,1,3-dimethoxy-5-bromobenzene,1-bromo-3,5-dimethoxy-benzene,1-bromo-3,5-dimethox,3,5-dimethoxy-bromobenzene,bromo-3,5-dimethoxybenzene,3,5-dimethoxy bromobenzene,acmc-1cs59,ksc495e7l
639187
1-bromo-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)Br)OC
20469-65-2
C8H9BrO2
217.06
KRWRFIMBWRVMKE-UHFFFAOYSA-N
3,5-dimethoxybromobenzene,benzene, 1-bromo-3,5-dimethoxy,1,3-dimethoxy-5-bromobenzene,1-bromo-3,5-dimethoxy-benzene,1-bromo-3,5-dimethox,3,5-dimethoxy-bromobenzene,bromo-3,5-dimethoxybenzene,3,5-dimethoxy bromobenzene,acmc-1cs59,ksc495e7l
639187
1-bromo-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)Br)OC
Melting Point 63.6°C to 65.7°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 217.06
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1-Bromo-3, 5-dimethoxybenzene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning