Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Bromo-3,5-dinitrobenzene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | White |
Quantity | 1 g |
UN Number | UN3459 |
Beilstein | 2053146 |
Synonym | 1-bromo-3,5-dinitro-benzene, benzene, 1-bromo-3,5-dinitro, benzene,1-bromo-3,5-dinitro, 3,5-dinitrobromobenzene, 1-bromo-3,5-dinitro benzene, 5-bromo-1,3-dinitrobenzene, bromodinitrobenzene, pubchem21414, acmc-1b5tp, 3-bromo 1,5-dinitrobenzene |
Solubility Information | Slightly soluble in water. |
Formula Weight | 247 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 1-Bromo-3,5-dinitrobenzene |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction