Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-3-chloro-2-iodobenzene, 97%

Click to view available options
Quantity:
250mg
1g
5g
450412-28-9
C6H3BrClI
317.348
MFCD09907935
GIMVCZMZRZGDTL-UHFFFAOYSA-N
2-bromo-6-chloroiodobenzene,1-bromo-2-iodo-3-chlorobenzene,benzene, 1-?bromo-?3-?chloro-?2-?iodo
10335963
1-bromo-3-chloro-2-iodobenzene
C1=CC(=C(C(=C1)Br)I)Cl
450412-28-9
C6H3BrClI
317.348
MFCD09907935
GIMVCZMZRZGDTL-UHFFFAOYSA-N
2-bromo-6-chloroiodobenzene,1-bromo-2-iodo-3-chlorobenzene,benzene, 1-?bromo-?3-?chloro-?2-?iodo
10335963
1-bromo-3-chloro-2-iodobenzene
C1=CC(=C(C(=C1)Br)I)Cl
Quantity 1g
Sensitivity Light Sensitive
Beilstein 9119447
Formula Weight 317.35
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-Bromo-3-chloro-2-iodobenzene