Alfa Aesar™ 1-Bromo-3-chloropropane, 99%

Manufacturer: Alfa Aesar™ A1139536

Quantity 500g

 View more versions of this product

Catalog No. AAA1139536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-70-6
    Molecular Formula Cl(CH2)3Br
    Formula Weight 157.44
    MDL Number MFCD00000998
    UN Number UN2688
    Beilstein 605278
    Synonym Trimethylene bromochloride
    Chemical Name or Material 1-Bromo-3-chloropropane

    Specifications

    Formula Weight 157.44
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 109-70-6
    Synonym Trimethylene bromochloride
    MDL Number MFCD00000998
    Melting Point −59°C
    Quantity 500g
    Flash Point >140°C (284°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 1-BROMO-3-CHLOROPROPANE
    RTECS Number TX4113000
    TSCA TSCA
    Molecular Formula Cl(CH2)3Br
    UN Number UN2688
    Boiling Point 142° to 145°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.
    Beilstein 605278
    Chemical Name or Material 1-Bromo-3-chloropropane
    Assay 99%

    Description

    Applications
    1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.

    Solubility
    Miscible with alcohols, chlorofom and ether. Immiscible with water.

    Notes
    Incompatible with strong oxidizing, reducing agents, amines, nitrides, azo/diazo compounds, alkali metals and epoxides.

    GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.

    GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit