Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-3-(trifluoromethoxy)benzene, 98+%

Click to view available options
Quantity:
1g
5g
25g
2252-44-0
C7H4BrF3O
241.007
MFCD00040943
WVUDHWBCPSXAFN-UHFFFAOYSA-N
1-bromo-3-trifluoromethoxy benzene,3-trifluoromethoxy bromobenzene,3-bromo trifluoromethoxy benzene,m-bromo trifluoromethoxy benzene,3-bromo-1-trifluoromethoxy benzene,benzene, 1-bromo-3-trifluoromethoxy,3-bromo-alpha,alpha,alpha-trifluoroanisole,bromo-3-trifluoromethoxy benzene,buttpark 91\57-27,1-bromo-3-trifluoromethoxy-benzene
519964
1-bromo-3-(trifluoromethoxy)benzene
C1=CC(=CC(=C1)Br)OC(F)(F)F
2252-44-0
C7H4BrF3O
241.007
MFCD00040943
WVUDHWBCPSXAFN-UHFFFAOYSA-N
1-bromo-3-trifluoromethoxy benzene,3-trifluoromethoxy bromobenzene,3-bromo trifluoromethoxy benzene,m-bromo trifluoromethoxy benzene,3-bromo-1-trifluoromethoxy benzene,benzene, 1-bromo-3-trifluoromethoxy,3-bromo-alpha,alpha,alpha-trifluoroanisole,bromo-3-trifluoromethoxy benzene,buttpark 91\57-27,1-bromo-3-trifluoromethoxy-benzene
519964
1-bromo-3-(trifluoromethoxy)benzene
C1=CC(=CC(=C1)Br)OC(F)(F)F
Refractive Index 1.462
Density 1.62
Boiling Point 156°C to 158°C
Quantity 1g
Flash Point 62°C (143°F)
Beilstein 1945938
Formula Weight 241.01
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-3-(trifluoromethoxy)benzene

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures