Alfa Aesar™ 1-Bromo-4-chlorobenzene, 98+%

Manufacturer: Alfa Aesar™ A1216030

Quantity 250g

 View more versions of this product

Catalog No. AAA1216030

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 106-39-8
    Molecular Formula C6H4BrCl
    Formula Weight 191.46
    MDL Number MFCD00000600
    UN Number UN2811
    Beilstein 1099014
    Synonym 4-Bromochlorobenzene
    Chemical Name or Material 1-Bromo-4-chlorobenzene

    Specifications

    Formula Weight 191.46
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Synonym 4-Bromochlorobenzene
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number CY9006000
    CAS 106-39-8
    Boiling Point 196°C
    Flash Point >100°C (212°F)
    Molecular Formula C6H4BrCl
    MDL Number MFCD00000600
    Melting Point 63° to 67°C
    Beilstein 1099014
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    UN Number UN2811
    Odor Aromatic
    Health Hazard 1 Danger
    Chemical Name or Material 1-Bromo-4-chlorobenzene
    Assay ≥98%

    Description

    Applications
    1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

    Solubility
    Soluble in water 0.045g/L.

    Notes
    Store in cool, dry place, in well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit