Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-(difluoromethyl)benzene, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
51776-71-7
C7H5BrF2
207.02
MFCD06657964
HUSPSWKWFREKSS-UHFFFAOYSA-N
1-bromo-4-difluoromethyl benzene,1-bromo-4-difluoromethyl-benzene,benzene, 1-bromo-4-difluoromethyl,4-bromo-alpha,alpha-difluorotoluene,4-bromobenzal fluoride,pubchem23296,acmc-1aukw,1-bromo-4-difluoro-methyl-benzene
2761183
1-bromo-4-(difluoromethyl)benzene
C1=CC(=CC=C1C(F)F)Br
51776-71-7
C7H5BrF2
207.02
MFCD06657964
HUSPSWKWFREKSS-UHFFFAOYSA-N
1-bromo-4-difluoromethyl benzene,1-bromo-4-difluoromethyl-benzene,benzene, 1-bromo-4-difluoromethyl,4-bromo-alpha,alpha-difluorotoluene,4-bromobenzal fluoride,pubchem23296,acmc-1aukw,1-bromo-4-difluoro-methyl-benzene
2761183
1-bromo-4-(difluoromethyl)benzene
C1=CC(=CC=C1C(F)F)Br
Infrared Spectrum Authentic
Boiling Point 74.0°C to 76.0°C (12.0 Torr)
Quantity 5g
Packaging Glass bottle
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 207.02
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(difluoromethyl)benzene
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning