Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-fluoro-2-iodobenzene, 98+%

Click to view available options
Quantity:
1g
5g
25g
202865-72-3
C6H3BrFI
300.897
MFCD00042578
QDCBSSUCEUMLGC-UHFFFAOYSA-N
2-bromo-5-fluoroiodobenzene,4-fluoro-2-iodobromobenzene,2-iodo-4-fluorobromobenzene,1-bromo-4-fluoro-2-iodo-benzene,benzene, 1-bromo-4-fluoro-2-iodo,2-bromo-5-fluoro-1-iodobenzene,pubchem3233,acmc-1cljm,ksc493k3j,1-bromo-2-iodo-4-fluorobenzene
2773370
1-bromo-4-fluoro-2-iodobenzene
C1=CC(=C(C=C1F)I)Br
202865-72-3
C6H3BrFI
300.897
MFCD00042578
QDCBSSUCEUMLGC-UHFFFAOYSA-N
2-bromo-5-fluoroiodobenzene,4-fluoro-2-iodobromobenzene,2-iodo-4-fluorobromobenzene,1-bromo-4-fluoro-2-iodo-benzene,benzene, 1-bromo-4-fluoro-2-iodo,2-bromo-5-fluoro-1-iodobenzene,pubchem3233,acmc-1cljm,ksc493k3j,1-bromo-2-iodo-4-fluorobenzene
2773370
1-bromo-4-fluoro-2-iodobenzene
C1=CC(=C(C=C1F)I)Br
Flash Point >110°C (230°F)
Quantity 5g
Boiling Point 235°C
Formula Weight 300.9
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-4-fluoro-2-iodobenzene

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning