Alfa Aesar™ 1-Bromo-4-fluorobenzene, 99%

Manufacturer: Alfa Aesar™ A1163022

Quantity 100g

 View more versions of this product

Catalog No. AAA1163022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 460-00-4
    Molecular Formula C6H4BrF
    Formula Weight 175.01
    MDL Number MFCD00000342
    UN Number UN1993
    Beilstein 774102
    Synonym 4-Bromofluorobenzene
    Chemical Name or Material 1-Bromo-4-fluorobenzene

    Specifications

    Formula Weight 175.01
    Molecular Formula C6H4BrF
    UN Number UN1993
    Health Hazard 3 GHS P Statement
    P261-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Synonym 4-Bromofluorobenzene
    RTECS Number CY9033750
    Melting Point −16°C
    Odor Aromatic
    CAS 460-00-4
    Boiling Point 151° to 153°C
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H226-H332-H319
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 60°C (140°F)
    MDL Number MFCD00000342
    Beilstein 774102
    Health Hazard 1 Warning
    TSCA TSCA
    Chemical Name or Material 1-Bromo-4-fluorobenzene
    Assay 99%

    Description

    Applications
    1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

    Solubility
    Soluble in water(0.14g/L).

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H226-H332-H319
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.

    GHS P Statement
    P261-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit