Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-fluorobutane, 97%

Click to view available options
Quantity:
1g
5g
25g
462-72-6
C4H8BrF
155.01
MFCD00209551
WMCUHRDQSHQNRW-UHFFFAOYSA-N
4-fluorobutyl bromide,butane, 1-bromo-4-fluoro,butane,1-bromo-4-fluoro,fluorobutyl bromide,acmc-20aoww,4-bromo-1-fluorobutane,1-bromo-4-fluoro-butane,4-01-00-00263 beilstein handbook reference,1-bromo-4-fluorobutane,1-bromo-4-fluoro-butan;4-fluoro-1-bromobutane
10022
1-bromo-4-fluorobutane
C(CCBr)CF
462-72-6
C4H8BrF
155.01
MFCD00209551
WMCUHRDQSHQNRW-UHFFFAOYSA-N
4-fluorobutyl bromide,butane, 1-bromo-4-fluoro,butane,1-bromo-4-fluoro,fluorobutyl bromide,acmc-20aoww,4-bromo-1-fluorobutane,1-bromo-4-fluoro-butane,4-01-00-00263 beilstein handbook reference,1-bromo-4-fluorobutane,1-bromo-4-fluoro-butan;4-fluoro-1-bromobutane
10022
1-bromo-4-fluorobutane
C(CCBr)CF
Applications
1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

Solubility
Sparingly soluble in water at 25°C 0.48 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 25g
Color Colorless
Refractive Index 1.4390
Density 1.444g/mL
Boiling Point 134°C to 135°C
Linear Formula FCH2CH2CH2CH2Br
UN Number UN1993
Beilstein 1731414
Formula Weight 155.01
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-fluorobutane
Show More Show Less

GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : EJ6229000