Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Bromo-4-iodobenzene, 98%, ACROS Organics™
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Quantity | 100g |
Color | White to Brown |
Infrared Spectrum | Authentic |
Melting Point | 89.0°C to 93.0°C |
Boiling Point | 120.0°C to 122.0°C (14.0 mmHg) |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | BrC6H4I |
Beilstein | 05,223 |
Formula Weight | 282.9 |
Show More |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 209-662-7