Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-iodonaphthalene, 98%

Click to view available options
Quantity:
250mg
1g
5g
63279-58-3
C10H6BrI
332.966
MFCD01166242
HQHHKYXPFKHLBF-UHFFFAOYSA-N
1-iodo-4-bromonaphthalene,1-iodo-4-bromoaphthalene,1-bromo-4-iodonaphth,naphthalene, 1-bromo-4-iodo
5073667
1-bromo-4-iodonaphthalene
C1=CC=C2C(=C1)C(=CC=C2I)Br
63279-58-3
C10H6BrI
332.966
MFCD01166242
HQHHKYXPFKHLBF-UHFFFAOYSA-N
1-iodo-4-bromonaphthalene,1-iodo-4-bromoaphthalene,1-bromo-4-iodonaphth,naphthalene, 1-bromo-4-iodo
5073667
1-bromo-4-iodonaphthalene
C1=CC=C2C(=C1)C(=CC=C2I)Br
Applications
It is used as an OLED material.

Solubility
Slightly soluble in water. (3.6 g/L) (25°C)

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 250mg
Sensitivity Light Sensitive
Melting Point 84°C to 89°C
Beilstein 2044950
Formula Weight 332.97
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-iodonaphthalene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning