Alfa Aesar™ 1-Bromo-4-methylnaphthalene, 98%

Manufacturer: Alfa Aesar™ H5503906

Quantity 5g

 View more versions of this product

Catalog No. AAH5503906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6627-78-7
    Molecular Formula C11H9Br
    Formula Weight 221.1
    MDL Number MFCD00003872
    Chemical Name or Material 1-Bromo-4-methylnaphthalene

    Specifications

    Formula Weight 221.1
    Boiling Point 162° to 164°C (12mmHg)
    Molecular Formula C11H9Br
    MDL Number MFCD00003872
    Quantity 5g
    Flash Point >113°C (235°F)
    CAS 6627-78-7
    Chemical Name or Material 1-Bromo-4-methylnaphthalene
    Assay 98%

    Description

    Applications
    It is used as a pharmaceutical intermediate.

    Solubility
    Sparingly Soluble in water. (1.9E-3 g/L) (25°C).

    Notes
    Store at room temperature with proper ventilation. Keep away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit