Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-methylnaphthalene, 98%

Manufacturer:  Alfa Aesar™ H5503914

Quantity 25g

 View more versions of this product

Catalog No. AAH5503914


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Sparingly Soluble in water. (1.9E-3 g/L) (25°C).

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

6627-78-7
221.1
1-Bromo-4-methylnaphthalene
C11H9Br
MFCD00003872

Specifications

6627-78-7
221.1
162° to 164°C (12mmHg)
>113°C (235°F)
98%
C11H9Br
MFCD00003872
25g
1-Bromo-4-methylnaphthalene
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit