Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-n-butylbenzene, 98%

Click to view available options
Quantity:
25g
100g
500g
41492-05-1
C10H13Br
213.118
MFCD00040934
BRGVKVZXDWGJBX-UHFFFAOYSA-N
1-bromo-4-n-butylbenzene,4-bromo-n-butylbenzene,4-n-butylbromobenzene,1-4-bromophenyl butane,benzene, 1-bromo-4-butyl,4-butyl bromobenzene,4-butyl-bromobenzene,4-1-butylbromobenzene,4-butyl phenylbromide,pubchem15109
521059
1-bromo-4-butylbenzene
CCCCC1=CC=C(C=C1)Br
41492-05-1
C10H13Br
213.118
MFCD00040934
BRGVKVZXDWGJBX-UHFFFAOYSA-N
1-bromo-4-n-butylbenzene,4-bromo-n-butylbenzene,4-n-butylbromobenzene,1-4-bromophenyl butane,benzene, 1-bromo-4-butyl,4-butyl bromobenzene,4-butyl-bromobenzene,4-1-butylbromobenzene,4-butyl phenylbromide,pubchem15109
521059
1-bromo-4-butylbenzene
CCCCC1=CC=C(C=C1)Br
Quantity 25g
Boiling Point 242°C
Flash Point 108°C (226°F)
Beilstein 2080263
Formula Weight 213.12
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-n-butylbenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning