Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-n-decylbenzene, 95%

Click to view available options
Quantity:
1g
5g
25g
106418-67-1
C16H25Br
297.28
MFCD00061112
SWTGVSCSSDYINB-UHFFFAOYSA-N
1-bromo-4-n-decylbenzene,benzene, 1-bromo-4-decyl,4-decylbromobenzene,acmc-2098jm,ksc909c2n
13757384
1-bromo-4-decylbenzene
CCCCCCCCCCC1=CC=C(C=C1)Br
106418-67-1
C16H25Br
297.28
MFCD00061112
SWTGVSCSSDYINB-UHFFFAOYSA-N
1-bromo-4-n-decylbenzene,benzene, 1-bromo-4-decyl,4-decylbromobenzene,acmc-2098jm,ksc909c2n
13757384
1-bromo-4-decylbenzene
CCCCCCCCCCC1=CC=C(C=C1)Br
Color Colorless to Yellow
Density 1.102g/mL
Quantity 25g
Refractive Index 1.5110
Beilstein 4743413
Formula Weight 297.28
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1-Bromo-4-n-decylbenzene