Alfa Aesar™ 1-Bromo-4-n-dodecylbenzene, 98%

Manufacturer: Alfa Aesar™ H3296303

Quantity 1g

 View more versions of this product

Catalog No. AAH3296303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 126930-72-1
    Molecular Formula C18H29Br
    Formula Weight 325.33
    MDL Number MFCD00191386
    Synonym 4-n-Decylbromobenzene; 4-n-Decylphenyl bromide
    Chemical Name or Material 1-Bromo-4-n-dodecylbenzene

    Specifications

    Formula Weight 325.33
    MDL Number MFCD00191386
    Quantity 1g
    Flash Point >110°C (230°F)
    Molecular Formula C18H29Br
    CAS 126930-72-1
    Synonym 4-n-Decylbromobenzene; 4-n-Decylphenyl bromide
    Boiling Point 376°C
    Chemical Name or Material 1-Bromo-4-n-dodecylbenzene
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit