Alfa Aesar™ 1-Bromo-4-n-heptylbenzene, 97%

Manufacturer: Alfa Aesar™ A1223503

Quantity 1g

 View more versions of this product

Catalog No. AAA1223503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76287-49-5
    Molecular Formula C13H19Br
    Formula Weight 255.2
    MDL Number MFCD00061115
    Synonym 4-n-Heptylbromobenzene
    Chemical Name or Material 1-Bromo-4-n-heptylbenzene

    Specifications

    Formula Weight 255.2
    MDL Number MFCD00061115
    Synonym 4-n-Heptylbromobenzene
    Quantity 1g
    TSCA TSCA
    Molecular Formula C13H19Br
    CAS 76287-49-5
    Chemical Name or Material 1-Bromo-4-n-heptylbenzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit