Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-n-heptyloxybenzene, 97%

Click to view available options
Quantity:
1g
5g
123732-04-7
C13H19BrO
271.198
MFCD00173752
DSCVEUSJTAFLBF-UHFFFAOYSA-N
1-4-bromophenoxy heptane,1-bromo-4-heptyloxybenzene,1-bromo-4-heptyloxy benzene,4-n-heptyloxybromobenzene,1-bromo-4-n-heptyloxybenzene,4-bromophenylheptyl ether,4-bromo-heptyloxy benzene
599102
1-bromo-4-heptoxybenzene
CCCCCCCOC1=CC=C(C=C1)Br
123732-04-7
C13H19BrO
271.198
MFCD00173752
DSCVEUSJTAFLBF-UHFFFAOYSA-N
1-4-bromophenoxy heptane,1-bromo-4-heptyloxybenzene,1-bromo-4-heptyloxy benzene,4-n-heptyloxybromobenzene,1-bromo-4-n-heptyloxybenzene,4-bromophenylheptyl ether,4-bromo-heptyloxy benzene
599102
1-bromo-4-heptoxybenzene
CCCCCCCOC1=CC=C(C=C1)Br
Quantity 1g
Formula Weight 271.2
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-n-heptyloxybenzene