Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-nitrobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
Plastic bottle
586-78-7
C6H4BrNO2
202.01
MFCD00007280
ZDFBKZUDCQQKAC-UHFFFAOYSA-N
4-bromonitrobenzene,4-nitrobromobenzene,p-bromonitrobenzene,benzene, 1-bromo-4-nitro,p-nitrobromobenzene,4-nitrophenyl bromide,4-bromo-1-nitrobenzene,p-nitrophenyl bromide,1-bromo-4-nitro-benzene,ccris 3115
11466
1-bromo-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])Br
586-78-7
C6H4BrNO2
202.01
MFCD00007280
ZDFBKZUDCQQKAC-UHFFFAOYSA-N
4-bromonitrobenzene,4-nitrobromobenzene,p-bromonitrobenzene,benzene, 1-bromo-4-nitro,p-nitrobromobenzene,4-nitrophenyl bromide,4-bromo-1-nitrobenzene,p-nitrophenyl bromide,1-bromo-4-nitro-benzene,ccris 3115
11466
1-bromo-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])Br
Quantity 10g
Packaging Glass bottle
Boiling Point 255.0°C to 256.0°C
Color Yellow
Melting Point 125.0°C to 127.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H4NO2
Beilstein 05, 248
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 202.01
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-nitrobenzene, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning

EINECSNumber : 209-583-8

RTECSNumber : CY9040550

TSCA : TSCA