Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-4-phenylbutane, 98%

Click to view available options
Quantity:
1g
5g
25g
13633-25-5
C10H13Br
213.118
MFCD00154988
XPBQQAHIVODAIC-UHFFFAOYSA-N
1-bromo-4-phenylbutane,4-bromobutyl benzene,4-phenylbutyl bromide,4-phenyl 1-butyl bromide,4-phenyl-1-butyl bromide,4-phenyl-1-bromobutane,1-bromo-4-phenyl-butane,benzene, 4-bromobutyl,benzene, bromobutyl,4-phenyl-1-bromo butane
259668
4-bromobutylbenzene
C1=CC=C(C=C1)CCCCBr
13633-25-5
C10H13Br
213.118
MFCD00154988
XPBQQAHIVODAIC-UHFFFAOYSA-N
1-bromo-4-phenylbutane,4-bromobutyl benzene,4-phenylbutyl bromide,4-phenyl 1-butyl bromide,4-phenyl-1-butyl bromide,4-phenyl-1-bromobutane,1-bromo-4-phenyl-butane,benzene, 4-bromobutyl,benzene, bromobutyl,4-phenyl-1-bromo butane
259668
4-bromobutylbenzene
C1=CC=C(C=C1)CCCCBr
Applications
1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Boiling Point 131°C to 133°C (12mmHg)
Quantity 25g
Formula Weight 213.12
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-phenylbutane