Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromo-5-fluoropentane, 99%

Click to view available options
Quantity:
250mg
1g
5g
407-97-6
C5H10BrF
169.037
MFCD01709395
GMYIZICPHREVDH-UHFFFAOYSA-N
pentane, 1-bromo-5-fluoro,5-fluoroamyl bromide,5-fluoropentyl bromide,fluoropentyl bromide,5-bromopentyl fluoride,acmc-20a2uo,1-bromo-5-fluoro-pentane,pentane,1-bromo-5-fluoro,4-01-00-00313 beilstein handbook reference,ksc493m6b
120236
1-bromo-5-fluoropentane
C(CCF)CCBr
407-97-6
C5H10BrF
169.037
MFCD01709395
GMYIZICPHREVDH-UHFFFAOYSA-N
pentane, 1-bromo-5-fluoro,5-fluoroamyl bromide,5-fluoropentyl bromide,fluoropentyl bromide,5-bromopentyl fluoride,acmc-20a2uo,1-bromo-5-fluoro-pentane,pentane,1-bromo-5-fluoro,4-01-00-00313 beilstein handbook reference,ksc493m6b
120236
1-bromo-5-fluoropentane
C(CCF)CCBr
Applications
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as μ-opioid receptor imaging agents.

Solubility
Soluble in methanol (in methanol). Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Refractive Index 1.4406
Boiling Point 162°C
Color Colorless
Quantity 5g
Density 1.360g/mL
Linear Formula F(CH2)5Br
Beilstein 1697258
Formula Weight 169.04
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Bromo-5-fluoropentane
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RZ9810000