Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-5-phenylpentane, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
14469-83-1
C11H15Br
227.14
MFCD01075177
QICUPOFVENZWSC-UHFFFAOYSA-N
5-bromopentyl benzene,1-bromo-5-phenylpentane,benzene, 5-bromopentyl,5-phenylpentyl bromide,1-bromo-5-phenyl-pentane,5-bromo-n-pentyl benzene,5-bromo-pentyl-benzene,5-brompentyl benzol,phenylpentyl bromide,5-phenylpentylbromide
285561
5-bromopentylbenzene
C1=CC=C(C=C1)CCCCCBr
14469-83-1
C11H15Br
227.14
MFCD01075177
QICUPOFVENZWSC-UHFFFAOYSA-N
5-bromopentyl benzene,1-bromo-5-phenylpentane,benzene, 5-bromopentyl,5-phenylpentyl bromide,1-bromo-5-phenyl-pentane,5-bromo-n-pentyl benzene,5-bromo-pentyl-benzene,5-brompentyl benzol,phenylpentyl bromide,5-phenylpentylbromide
285561
5-bromopentylbenzene
C1=CC=C(C=C1)CCCCCBr
Boiling Point 148°C (18.0mmHg)
Specific Gravity 1.23
Density 1.2300g/mL
Refractive Index 1.5320 to 1.534
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5(CH2)5Br
Formula Weight 227.14
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Bromo-5-phenylpentane
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.