Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromoadamantane, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
768-90-1
C10H15Br
215.13
MFCD00074721
VQHPRVYDKRESCL-UHFFFAOYSA-N
1-adamantyl bromide,adamantyl bromide,bromoadamantane,1-bromo-adamantane,adamantane, 1-bromo,1-bromotricyclo 3.3.1.13,7 decane,tricyclo 3.3.1.13,7 decane, 1-bromo,tricyclo 3.3.1.1 3,7-decane, 1-bromo,labotest-bb lt00239617,3r,5s,7s-1-bromoadamantane
79106
1-bromoadamantane
C1C2CC3CC1CC(C2)(C3)Br
768-90-1
C10H15Br
215.13
MFCD00074721
VQHPRVYDKRESCL-UHFFFAOYSA-N
1-adamantyl bromide,adamantyl bromide,bromoadamantane,1-bromo-adamantane,adamantane, 1-bromo,1-bromotricyclo 3.3.1.13,7 decane,tricyclo 3.3.1.13,7 decane, 1-bromo,tricyclo 3.3.1.1 3,7-decane, 1-bromo,labotest-bb lt00239617,3r,5s,7s-1-bromoadamantane
79106
1-bromoadamantane
C1C2CC3CC1CC(C2)(C3)Br
Color White to Yellow
Packaging Glass bottle
Quantity 25g
Melting Point 117.0°C to 120.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,chloroform,benzene,,methanol and ethanol
Formula Weight 215.13
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 1-Bromoadamantane, 99%
Show More Show Less

MOISTURE SENSITIVE

EINECSNumber : 212-200-7

TSCA : TSCA