Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromobutane, 98%, Spectrum™

Click to view available options
Quantity:
25 g
250 g
1 kg
2.5 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
109-65-9
109-65-9
Packaging Poly-Coated Glass Bottle
Quantity 2.5 kg
CAS Max % 1
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.