Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromohexadecane, 97%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
112-82-3
C16H33Br
305.34
MFCD00000230
HNTGIJLWHDPAFN-UHFFFAOYSA-N
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
8213
1-bromohexadecane
CCCCCCCCCCCCCCCCBr
112-82-3
C16H33Br
305.34
MFCD00000230
HNTGIJLWHDPAFN-UHFFFAOYSA-N
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
8213
1-bromohexadecane
CCCCCCCCCCCCCCCCBr
Specific Gravity 0.999
Density 0.9990g/mL
Color Amber to Black-Brown
Flash Point 177°C
Packaging Glass bottle
Quantity 2.5L
Refractive Index 1.4609 to 1.4629
Boiling Point 336.0°C
Melting Point 17.3°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)15Br
Beilstein 01,172
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 305.33
Physical Form Liquid After Melting
Percent Purity 97%
Chemical Name or Material 1-Bromohexadecane
Show More Show Less

EINECSNumber : 204-008-7