Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Bromomethyl)adamantane, 97%, Maybridge™

Click to view available options
Quantity:
5g
10g
Packaging:
Amber Glass Bottle
14651-42-4
C11H17Br
229.161
RLRBYCAVZBQANU-UHFFFAOYSA-N
1-bromomethyl adamantane, tricyclo 3.3.1.13,7 decane, 1-bromomethyl, adamantanylbromomethane, 1-bromomethyl tricyclo 3.3.1.1~3,7~ decane, 3r,5s,7s-1-bromomethyl adamantane, pubchem21524, 1-adamantylmethylbromide, 1-bromoethyl adamantane, 1-adamantyl methyl bromide, 1s,3r,5s,7s-1-bromomethyl adamantane
1416304
1-(bromomethyl)adamantane
C1C2CC3CC1CC(C2)(C3)CBr
14651-42-4
C11H17Br
229.161
RLRBYCAVZBQANU-UHFFFAOYSA-N
1-bromomethyl adamantane, tricyclo 3.3.1.13,7 decane, 1-bromomethyl, adamantanylbromomethane, 1-bromomethyl tricyclo 3.3.1.1~3,7~ decane, 3r,5s,7s-1-bromomethyl adamantane, pubchem21524, 1-adamantylmethylbromide, 1-bromoethyl adamantane, 1-adamantyl methyl bromide, 1s,3r,5s,7s-1-bromomethyl adamantane
1416304
1-(bromomethyl)adamantane
C1C2CC3CC1CC(C2)(C3)CBr
Quantity 10g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 229.16
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)adamantane