Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromooctadecane, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
112-89-0
C18H37Br
333.398
MFCD00000231
WSULSMOGMLRGKU-UHFFFAOYSA-N
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
8218
1-bromooctadecane
CCCCCCCCCCCCCCCCCCBr
112-89-0
C18H37Br
333.398
MFCD00000231
WSULSMOGMLRGKU-UHFFFAOYSA-N
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
8218
1-bromooctadecane
CCCCCCCCCCCCCCCCCCBr
Packaging Glass Bottle
Infrared Spectrum Authentic
Refractive Index 1.4620 to 1.4640
Flash Point >110°C
Specific Gravity 0.97
Quantity 5g
Melting Point >20.0°C
Density 0.9700g/mL
Boiling Point 214.0°C to 216.0°C (12.0mmHg)
Color Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CH3(CH2)17Br
Formula Weight 333.38
Physical Form Low Melting Solid
Percent Purity 96%
Chemical Name or Material 1-Bromooctadecane, 96%
Show More Show Less

EINECSNumber : 204-013-4

TSCA : TSCA