Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromooctadecane, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
112-89-0
C18H37Br
333.38
MFCD00000231
WSULSMOGMLRGKU-UHFFFAOYSA-N
octadecyl bromide,stearyl bromide,octadecane, 1-bromo,n-octadecyl bromide,bromooctadecane,octadecylbromide,1-bromoctadecane,n-octadecylbromide,1-bromooctodecane,bromo-n-octadecane
8218
1-bromooctadecane
CCCCCCCCCCCCCCCCCCBr
112-89-0
C18H37Br
333.38
MFCD00000231
WSULSMOGMLRGKU-UHFFFAOYSA-N
octadecyl bromide,stearyl bromide,octadecane, 1-bromo,n-octadecyl bromide,bromooctadecane,octadecylbromide,1-bromoctadecane,n-octadecylbromide,1-bromooctodecane,bromo-n-octadecane
8218
1-bromooctadecane
CCCCCCCCCCCCCCCCCCBr
Density 0.9700g/mL
Specific Gravity 0.97
Boiling Point 214.0°C to 216.0°C (12.0mmHg)
Melting Point >20.0°C
Refractive Index 1.4620 to 1.4640
Quantity 100g
Color Yellow
Packaging Glass bottle
Flash Point >110°C
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula CH3(CH2)17Br
Beilstein 01,I,69
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 333.38
Physical Form Low Melting Solid
Percent Purity 96%
Chemical Name or Material 1-Bromooctadecane, 96%
Show More Show Less

EINECSNumber : 204-013-4

TSCA : TSCA