Alfa Aesar™ 1-Bromoperfluorooctane, 98+%

Manufacturer: Alfa Aesar™ B2247003

Quantity 1g

 View more versions of this product

Catalog No. AAB2247003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 423-55-2
    Molecular Formula C8H17Br
    Linear Formula CF3(CF2)7Br
    Formula Weight 498.98
    MDL Number MFCD00042082
    Beilstein 2016022
    Merck Index 14,7156
    Synonym Perflubron; Perfluorooctyl bromide
    Chemical Name or Material 1-Bromoperfluorooctane

    Specifications

    Formula Weight 498.98
    Boiling Point 141° to 143°C
    Quantity 1g
    Linear Formula CF3(CF2)7Br
    Beilstein 2016022
    Merck Index 14,7156
    Synonym Perflubron; Perfluorooctyl bromide
    Health Hazard 1 Warning
    Melting Point 6°C
    Molecular Formula C8H17Br
    MDL Number MFCD00042082
    CAS 423-55-2
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number RG8700000
    Chemical Name or Material 1-Bromoperfluorooctane
    Assay ≥98%

    Description

    Applications
    1-Bromoperfluorooctane 1-Bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier. It is used as an O2 carrier in some emulsions.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Incompatible with oxidizing agents, acids, metals.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit