Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Bromoperfluorooctane, 98+%

Click to view available options
Quantity:
1g
5g
25g
423-55-2
C8BrF17
498.965
MFCD00042082
WTWWXOGTJWMJHI-UHFFFAOYSA-N
perflubron,1-bromoperfluorooctane,perfluorooctyl bromide,1-bromoheptadecafluorooctane,oxygent,perfluoro-n-octyl bromide,imagent gi,perfluorooctylbromide,liquivent,perfluoroctyl bromide
9873
CHEBI:38803
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
C(C(C(C(C(F)(F)Br)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F
423-55-2
C8BrF17
498.965
MFCD00042082
WTWWXOGTJWMJHI-UHFFFAOYSA-N
perflubron,1-bromoperfluorooctane,perfluorooctyl bromide,1-bromoheptadecafluorooctane,oxygent,perfluoro-n-octyl bromide,imagent gi,perfluorooctylbromide,liquivent,perfluoroctyl bromide
9873
CHEBI:38803
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
C(C(C(C(C(F)(F)Br)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F
Applications
1-Bromoperfluorooctane 1-Bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier. It is used as an O2 carrier in some emulsions.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with oxidizing agents, acids, metals.
Boiling Point 141°C to 143°C
Melting Point 6°C
Quantity 5g
Linear Formula CF3(CF2)7Br
Beilstein 2016022
Merck Index 14,7156
Formula Weight 498.98
Percent Purity ≥98%
Chemical Name or Material 1-Bromoperfluorooctane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RG8700000